This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03021693_g1
See other TPI1 GE Assays ›
Gene Symbol:
Tpi1 LOC100911515
Gene Name:
triosephosphate isomerase 1
triosephosphate isomerase-like
Gene Aliases:
Tpi
-
Chromosome Location:
Chr.4: 157328375 - 157331905 on Build Rnor_6.0
Chromosome Location:
Chr.9: 20213596 - 20217278 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn03021693_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tpi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022922.2 NP_075211.2 7 1338 100
GenBank mRNA BC126087.1 - 7 1338 8368624 100
Gene Symbol: LOC100911515
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003750654.3 XP_003750702.2 7 1494 100

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tpi1
Click here to find siRNAs for Gene LOC100911515

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
gluconeogenesis
glycolytic process
pentose-phosphate shunt
glucose metabolic process
multicellular organism development
glyceraldehyde-3-phosphate metabolic process
triose-phosphate isomerase activity
protein binding
isomerase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065922 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P673370 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 468233 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 415360 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 40537 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065922 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P673370 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065922 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P673370 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065922 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P673370 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121160 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700146861 - -
Agilent Rat cDNA Microarray (G4105A) 700146861 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10798022 - -
Affymetrix Rat Gene 1.0 ST Array 10865529 - -
Affymetrix Rat Gene 1.0 ST Array 10865530 - -
Affymetrix Rat Gene 1.0 ST Array 10865528 - -
Affymetrix Rat Gene 1.1 ST Array 10865527 - -
Affymetrix Rat Gene 1.1 ST Array 10798021 - -
Rat Genome 230 2.0 Array 1367603_PM_at - -
Rat Genome 230 2.0 Array 1367603_at - -
Affymetrix HT Rat Focus Array 1367603_at - -
Affymetrix HT Rat Focus Array 1367603_PM_at - -
Rat Expression Set 230 1367603_PM_at - -
Rat Expression Set 230 1367603_at - -
Affymetrix Rat Gene 1.0 ST Array 10798021 - -
Affymetrix Rat Gene 1.0 ST Array 10865527 - -
Rat Genome U34 Set rc_AI104399_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21623507 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1650818 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0