Assay Details
| Gene Symbol: Vom2r16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099655.1 | NP_001093125.1 | 4 - 5 | 1524 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r16| Gene Symbol: Vom2r16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099655.1 | NP_001093125.1 | 4 - 5 | 1524 | 99 | |
Set Membership: |
|