This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03296716_s1
See other MT-ATP8 GE Assays ›
Gene Symbol:
MT-ATP8
Gene Name:
mitochondrially encoded ATP synthase 8
Gene Aliases:
ATP8
Chromosome Location:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03296716_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MT-ATP8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA NC_001665.ATP8.0 - 48 101

Back To Top

More Information


Additional Information:

NC_001665 (strand:+; 7758..7961); mitochondrially encoded ATP synthase 8

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mitochondria Encoded Recommended Assay Rattus norvegicus Inventoried

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Rat Genome U34 Set rc_AI179510_f_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0