This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03399815_s1
See other GPR146 GE Assays ›
Gene Symbol:
Gpr146
Gene Name:
G protein-coupled receptor 146
Gene Aliases:
RGD1560731
Chromosome Location:
Chr.12: 17343396 - 17358628 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03399815_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006248955.2 XP_006249017.1 2 5396 61
XM_006248956.2 XP_006249018.1 2 3730 61
XM_006248957.2 XP_006249019.1 2 3264 61
XM_006248958.2 XP_006249020.1 3 5004 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20714968 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline