This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03416077_m1
See other CEACAM6 GE Assays ›
Gene Symbol:
Ceacam6 LOC103689938
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 6
carcinoembryonic antigen-related cell adhesion molecule 1-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 81683500 - 81701230 on Build Rnor_6.0
Chromosome Location:
Chr.1: 83043478 - 83061421 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn03416077_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ceacam6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045097.1 5 - 6 1127 109
GenBank mRNA AB434131.1 - 4 - 5 955 109
BC078962.1 - 5 - 6 1127 7111028 109
Gene Symbol: LOC103689938
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008759078.1 XP_008757300.1 1237 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100001056.rn.1, TC0100001096.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088677 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 899925 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 214994 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 796025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391295 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 487981 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 86710 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 666356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 121094 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 251743 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 642060 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 329868 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 547107 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 907554 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 4540 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145730 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088675 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088675 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088675 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 96590 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10705038 - -
Affymetrix Rat Gene 1.0 ST Array 10705042 - -
Affymetrix Rat Gene 1.0 ST Array 10705046 - -
Affymetrix Rat Gene 1.0 ST Array 10705044 - -
Affymetrix Rat Gene 1.0 ST Array 10705039 - -
Affymetrix Rat Gene 1.0 ST Array 10705043 - -
Affymetrix Rat Gene 1.0 ST Array 10705048 - -
Affymetrix Rat Gene 1.0 ST Array 10705041 - -
Affymetrix Rat Gene 1.0 ST Array 10705040 - -
Affymetrix Rat Gene 1.0 ST Array 10705045 - -
Affymetrix Rat Gene 1.0 ST Array 10705047 - -
Affymetrix Rat Gene 1.1 ST Array 10705037 - -
Affymetrix Rat Clariom S Assay PSR0100007505.rn.1 TC0100001056.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100007972.rn.1 TC0100001096.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100007973.rn.1 TC0100001096.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10705037 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-jhckj:80/100.66.72.99:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0