This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03648126_g1
See other EEF1B2 GE Assays ›
Gene Symbol:
Eef1b2
Gene Name:
eukaryotic translation elongation factor 1 beta 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 69953440 - 69956158 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03648126_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108799.2 NP_001102269.1 5 - 6 823 136
GenBank mRNA BC168738.1 - 5 - 6 823 6918011 136
FQ216958.1 - 5 - 6 591 136
FQ217523.1 - 5 - 6 591 136
FQ218320.1 - 5 - 6 591 136
FQ221159.1 - 5 - 6 591 136
FQ221342.1 - 5 - 6 593 136
FQ222681.1 - 5 - 6 591 136
FQ222690.1 - 5 - 6 592 136
FQ223187.1 - 5 - 6 591 136
FQ228255.1 - 5 - 6 592 136
FQ228375.1 - 5 - 6 593 136
FQ228595.1 - 5 - 6 592 136
FQ229059.1 - 5 - 6 593 136
FQ232532.1 - 5 - 6 594 136

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
translation elongation factor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10457 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 928304 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 943634 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10457 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10457 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10457 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P263072 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10830104 - -
Affymetrix Rat Gene 1.0 ST Array 10923860 - -
Affymetrix Rat Gene 1.0 ST Array 10923861 - -
Affymetrix Rat Gene 1.0 ST Array 10923858 - -
Affymetrix Rat Gene 1.0 ST Array 10923859 - -
Affymetrix Rat Gene 1.1 ST Array 10830102 - -
Affymetrix Rat Gene 1.1 ST Array 10923857 - -
Rat Genome 230 2.0 Array 1388449_PM_at - -
Rat Genome 230 2.0 Array 1388449_at - -
Affymetrix HT Rat Focus Array 1388449_PM_at - -
Affymetrix HT Rat Focus Array 1388449_at - -
Affymetrix Rat Clariom S Assay PSR0900006380.rn.1 TC0900000789.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900006378.rn.1 TC0900000789.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900006379.rn.1 TC0900000789.rn.1 RTA TC
Rat Expression Set 230 1388449_at - -
Rat Expression Set 230 1388449_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10923857 - -
Rat Genome U34 Set rc_AI175551_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1350882 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0