This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04222807_gH
See other HIST1H2BD GE Assays ›
Gene Symbol:
Hist1h2bd Hist1h2bh LOC100359692
Gene Name:
histone cluster 1, H2bd
histone cluster 1, H2bh
histone cluster 1, H2bc-like
Gene Aliases:
Hist1h2bm
-
-
Chromosome Location:
Chr.17: 44833924 - 44834305 on Build Rnor_6.0
Chromosome Location:
Chr.17: 43679925 - 43689311 on Build Rnor_6.0
Chromosome Location:
Chr.17: 44519815 - 44520269 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
Assay ID Rn04222807_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hist1h2bd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108414.1 NP_001101884.1 1 147 87
Gene Symbol: Hist1h2bh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107352.1 NP_001100822.1 1 87
GenBank mRNA BC079458.1 - 1 87
Gene Symbol: LOC100359692
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006254120.2 XP_006254182.1 1 87

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

innate immune response in mucosa
antibacterial humoral response
defense response to Gram-positive bacterium
DNA binding
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014508 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014508 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014508 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014508 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P219921 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229528 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10798498 - -
Affymetrix Rat Gene 1.0 ST Array 10798496 - -
Affymetrix Rat Gene 1.0 ST Array 10798489 - -
Affymetrix Rat Gene 1.1 ST Array 10798501 - -
Affymetrix Rat Gene 1.1 ST Array 10798494 - -
Affymetrix Rat Gene 1.1 ST Array 10798497 - -
Rat Genome 230 2.0 Array 1391301_at - -
Rat Genome 230 2.0 Array 1391301_PM_at - -
Affymetrix Rat Clariom S Assay PSR1700012523.rn.1 TC1700001832.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700012387.rn.1 TC1700001802.rn.1 RTA TC
Rat Expression Set 230 1391301_PM_at - -
Rat Expression Set 230 1391301_at - -
Affymetrix Rat Gene 1.0 ST Array 10798494 - -
Affymetrix Rat Gene 1.0 ST Array 10798488 - -
Affymetrix Rat Gene 1.0 ST Array 10798497 - -
Rat Genome U34 Set rc_AA964055_at - -
Rat Genome U34 Set rc_AA997499_i_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20847880 - -
ABI Rat Genome Survey Microarray 21398673 - -
ABI Rat Genome Survey Microarray 21600012 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359983 - -
Illumina ratRef-12 v1.0 expression beadchip ILMN_1351946 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline