This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244833_m1
See other ROMO1 GE Assays ›
Gene Symbol:
Romo1 LOC100910944
Gene Name:
reactive oxygen species modulator 1
reactive oxygen species modulator 1-like
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 151688213 - 151689813 on Build Rnor_6.0
Chromosome Location:
Chr.3: 152222726 - 152224314 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn04244833_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Romo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195490.1 NP_001182419.1 2 - 3 198 85
GenBank mRNA CF108387.1 - 1 - 2 85
FQ211245.1 - 2 - 3 183 85
Gene Symbol: LOC100910944
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006235452.1 XP_006235514.1 2 - 3 198 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300002135.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Function(s)

replicative cell aging
positive regulation of cell proliferation
cellular response to reactive oxygen species
positive regulation of reactive oxygen species metabolic process

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011194 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108222 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011194 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108222 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011194 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108222 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P159745 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P159743 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1399004_at - -
Rat Genome 230 2.0 Array 1399004_PM_at - -
Affymetrix HT Rat Focus Array 1399004_at - -
Affymetrix HT Rat Focus Array 1399004_PM_at - -
Affymetrix Rat Clariom S Assay PSR0300015744.rn.1 TC0300002135.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300015748.rn.1 TC0300002135.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300015747.rn.1 TC0300002135.rn.1 RTA TC
Rat Expression Set 230 1399004_at - -
Rat Expression Set 230 1399004_PM_at - -
Rat Genome U34 Set rc_AI029733_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20941418 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0