This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04341822_m1
See other FABP7 GE Assays ›
Gene Symbol:
Fabp7
Gene Name:
fatty acid binding protein 7, brain
Gene Aliases:
-
Chromosome Location:
Chr.20: 40769624 - 40773156 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04341822_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fabp7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030832.2 NP_110459.1 2 - 3 333 86
GenBank mRNA U02096.1 - 2 - 3 334 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fabp7

Gene Ontology Categories:

Function(s) Process(es)

startle response
transport
cell proliferation in forebrain
neurogenesis
epithelial cell proliferation
prepulse inhibition
transporter activity
fatty acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0