This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04419054_g1
See other LOC100233176 GE Assays ›
Gene Symbol:
LOC100233176
Gene Name:
hypothetical protein LOC100233176
Gene Aliases:
-
Chromosome Location:
Chr.6: 126620597 - 126636484 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04419054_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100233176
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142942.2 NP_001136414.1 1 - 2 260 72
GenBank mRNA CB612989.1 - 1 - 2 265 72

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600021696.rn.1 TC0600003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600021697.rn.1 TC0600003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600021707.rn.1 TC0600003053.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600021708.rn.1 TC0600003053.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline