This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06151526_m1
See other UGT3A2 GE Assays ›
Gene Symbol:
Ugt3a2
Gene Name:
UDP glycosyltransferase 3 family, polypeptide A2
Gene Aliases:
RGD1564365
Chromosome Location:
Chr.2: 58666337 - 58682212 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06151526_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ugt3a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008760833.1 XP_008759055.1 2 - 3 549 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

flavonoid biosynthetic process
cellular response to hormone stimulus
flavonoid glucuronidation
UDP-glycosyltransferase activity
glucuronosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0