This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06155197_m1
See other RGD1311318 GE Assays ›
Gene Symbol:
RGD1311318 RGD1560162
Gene Name:
similar to Arylacetamide deacetylase (AADAC)
similar to hypothetical protein C130079G13
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 149969697 - 150022733 on Build Rnor_6.0
Chromosome Location:
Chr.2: 150057676 - 150081312 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06155197_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGD1311318
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_227185.4 XP_227185.3 608 109
Gene Symbol: RGD1560162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_578020.5 XP_578020.5 4 - 5 608 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline