This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06168162_s1
See other DSEL GE Assays ›
Gene Symbol:
Dsel
Gene Name:
dermatan sulfate epimerase-like
Gene Aliases:
RGD1307724
Chromosome Location:
Chr.13: 1942384 - 1948079 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06168162_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dsel
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006249608.2 XP_006249670.1 1 632 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006249608.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
chondroitin sulfate metabolic process
dermatan sulfate metabolic process
sulfotransferase activity
chondroitin-glucuronate 5-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0