This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06345935_m1
See other CD200R1L GE Assays ›
Gene Symbol:
Cd200r1l
Gene Name:
CD200 receptor 1-like
Gene Aliases:
Cd200r2, RGD1563387
Chromosome Location:
Chr.11: 60803856 - 60828981 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06345935_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008768755.1 XP_008766977.1 1 - 2 69 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008768755.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline