This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06379955_g1
See other NUPR1 GE Assays ›
Gene Symbol:
Nupr1 NEWGENE_69363
Gene Name:
nuclear protein, transcriptional regulator, 1
nuclear protein, transcriptional regulator, 1
Gene Aliases:
p8
Nupr1
Chromosome Location:
Chr.1: 194767484 - 194769519 on Build Rnor_6.0
Chromosome Location:
Chr.1: 198043117 - 198045203 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06379955_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nupr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053611.2 NP_446063.1 3 497 82
GenBank mRNA AF014503.1 - 3 465 82
FQ221101.1 - 3 459 82
Gene Symbol: NEWGENE_69363
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008759924.1 XP_008758146.1 3 548 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nupr1
Click here to find siRNAs for Gene NEWGENE_69363

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0