This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06383183_g1
See other EIF4EBP1 GE Assays ›
Gene Symbol:
Eif4ebp1
Gene Name:
eukaryotic translation initiation factor 4E binding protein 1
Gene Aliases:
PHAS-I
Chromosome Location:
Chr.16: 68954860 - 68968248 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06383183_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4ebp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053857.2 NP_446309.1 3 56
GenBank mRNA AI603218.1 - 1 208 56
U05014.1 - 3 56

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4ebp1

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
IRES-dependent translational initiation
response to ischemia
insulin receptor signaling pathway
lung development
negative regulation of protein complex assembly
TOR signaling
response to ethanol
positive regulation of mitotic cell cycle
negative regulation of translational initiation
eukaryotic initiation factor 4E binding
translation repressor activity
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0