This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06411322_s1
See other MLYCD GE Assays ›
Gene Symbol:
Mlycd
Gene Name:
malonyl-CoA decarboxylase
Gene Aliases:
Mcd
Chromosome Location:
Chr.19: 52032950 - 52048773 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06411322_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mlycd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053477.1 NP_445929.1 5 1791 86
GenBank mRNA AF304865.1 - 5 1793 86
AJ007704.1 - 5 1791 86
BC061845.1 - 5 1796 5621015 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mlycd

Gene Ontology Categories:

Function(s) Process(es)

response to ischemia
acetyl-CoA biosynthetic process
fatty acid biosynthetic process
regulation of glucose metabolic process
fatty acid oxidation
regulation of fatty acid beta-oxidation
regulation of fatty acid oxidation
positive regulation of fatty acid oxidation
malonyl-CoA catabolic process
receptor binding
malonyl-CoA decarboxylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0