This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06423449_s1
See other GJB4 GE Assays ›
Gene Symbol:
Gjb4
Gene Name:
gap junction protein, beta 4
Gene Aliases:
Cnx30.3, Cx30.3
Chromosome Location:
Chr.5: 145416343 - 145421038 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06423449_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053984.2 NP_446436.1 1 904 74
XM_008763985.1 XP_008762207.1 2 634 74
XM_008763986.1 XP_008762208.1 1 232 74
GenBank mRNA X76168.1 - 1 904 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gjb4

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

cell communication
sensory perception of smell
biological_process
olfactory behavior
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline