This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06666431_g1
See other EIF4E GE Assays ›
Gene Symbol:
Eif4e
Gene Name:
eukaryotic translation initiation factor 4E
Gene Aliases:
-
Chromosome Location:
Chr.2: 243819614 - 243853987 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06666431_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053974.2 NP_446426.1 7 1552 81
XM_008761488.1 XP_008759710.1 8 1665 81
GenBank mRNA BC087001.1 - 7 1552 7122606 81
X83399.1 - 7 1586 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4e

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
behavioral fear response
translation
translational initiation
regulation of translation
lung development
positive regulation of mitotic cell cycle
translation initiation factor activity
protein binding
eukaryotic initiation factor 4G binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0