This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07309329_g1
See other GNGT2 GE Assays ›
Gene Symbol:
Gngt2
Gene Name:
guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 83654325 - 83658601 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07309329_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gngt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135767.1 NP_001129239.1 3 - 4 285 94
XM_006247208.2 XP_006247270.1 3 - 4 396 94
XM_006247209.1 XP_006247271.1 3 - 4 194 94
XM_006247210.1 XP_006247272.1 3 - 4 194 94
XM_006247211.2 XP_006247273.1 2 - 3 444 94
XM_008768115.1 XP_008766337.1 2 - 3 341 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
biological_process
metabolic process
molecular_function
GTPase activity
signal transducer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline