Assay Details
| Gene Symbol: Tas2r129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166683.1 | NP_001160155.1 | 1 | 817 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r129| Gene Symbol: Tas2r129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166683.1 | NP_001160155.1 | 1 | 817 | 101 | |
Set Membership: |
|