This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07311877_g1
See other ARC GE Assays ›
Gene Symbol:
Arc
Gene Name:
activity-regulated cytoskeleton-associated protein
Gene Aliases:
rg3.1
Chromosome Location:
Chr.7: 115907097 - 115911054 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07311877_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019361.1 NP_062234.1 2 - 3 2146 68
GenBank mRNA U19866.1 - 2 - 3 2146 68
Z46925.1 - 2 - 3 2123 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arc

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cytoskeleton organization
endoderm development
learning
anterior/posterior pattern specification
cell migration
regulation of cell morphogenesis
regulation of neuronal synaptic plasticity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0