Assay Details
| Gene Symbol: Pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022533.1 | NP_071978.1 | 2 - 3 | 523 | 81 | |
| GenBank mRNA | BC078823.1 | - | 2 - 3 | 403 | 7108239 | 81 |
| U13617.1 | - | 2 - 3 | 296 | 81 | ||
| Z49858.1 | - | 2 - 3 | 523 | 81 | ||
More Information
Set Membership: |
|

Pllp
Genomic Map