This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07312699_g1
See other TMED2 GE Assays ›
Gene Symbol:
Tmed2
Gene Name:
transmembrane emp24 domain trafficking protein 2
Gene Aliases:
Rnp24
Chromosome Location:
Chr.12: 37471259 - 37480346 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07312699_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmed2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031722.2 NP_113910.1 2 - 3 425 76
GenBank mRNA BC062036.1 - 2 - 3 425 5623841 76
X92097.1 - 2 - 3 396 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmed2

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
somitogenesis
neural tube closure
embryonic placenta development
maternal placenta development
heart looping
intracellular protein transport
vesicle targeting
Golgi organization
positive regulation of gene expression
somite rostral/caudal axis specification
negative regulation of GTPase activity
multicellular organism growth
post-anal tail morphogenesis
embryonic morphogenesis
protein targeting to plasma membrane
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0