This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07313292_m1
See other DHRS9 GE Assays ›
Gene Symbol:
Dhrs9
Gene Name:
dehydrogenase/reductase (SDR family) member 9
Gene Aliases:
Rdhl
Chromosome Location:
Chr.3: 55623671 - 55645883 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07313292_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130819.1 NP_570832.1 1 - 2 152 71
GenBank mRNA AF337953.1 - 1 - 2 152 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF337953.1, NM_130819.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhrs9

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

retinoic acid biosynthetic process
androgen metabolic process
progesterone metabolic process
retinoic acid metabolic process
9-cis-retinoic acid biosynthetic process
oxidation-reduction process
alcohol dehydrogenase (NAD) activity
retinol dehydrogenase activity
testosterone dehydrogenase (NAD+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline