This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07313593_m1
See other GRASP GE Assays ›
Gene Symbol:
Grasp
Gene Name:
GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 142869785 - 142877551 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07313593_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grasp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138894.1 NP_620249.1 6 - 7 667 77
GenBank mRNA AF374272.1 - 6 - 7 667 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grasp

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein localization
protein binding
PDZ domain binding
ADP-ribosylation factor binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hvxbk:80/100.66.77.65:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0