This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07317794_s1
See other GPR75 GE Assays ›
Gene Symbol:
Gpr75
Gene Name:
G protein-coupled receptor 75
Gene Aliases:
-
Chromosome Location:
Chr.14: 115270771 - 115278510 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07317794_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109096.1 NP_001102566.1 1 1522 72
XM_006251616.2 XP_006251678.1 1 1627 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
regulation of neuron death
G-protein coupled receptor activity
C-C chemokine receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline