This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07318645_s1
See other CXXC5 GE Assays ›
Gene Symbol:
Cxxc5
Gene Name:
CXXC finger protein 5
Gene Aliases:
-
Chromosome Location:
Chr.18: 28653653 - 28656908 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07318645_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cxxc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007628.2 NP_001007629.1 2 1221 96
GenBank mRNA DV723039.1 - 1 104 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cxxc5

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0