This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07365155_m1
See other IP6K3 GE Assays ›
Gene Symbol:
Ip6k3
Gene Name:
inositol hexakisphosphate kinase 3
Gene Aliases:
Ihpk3
Chromosome Location:
Chr.20: 5733640 - 5746553 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07365155_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ip6k3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008772778.1 XP_008771000.1 4 - 5 1340 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
inositol phosphate biosynthetic process
phosphatidylinositol metabolic process
inositol hexakisphosphate 6-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline