This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04109552_s1
See other TPI1 GE Assays ›
Gene Symbol:
TPI1
Gene Name:
Gene Aliases:
YDR050C
Chromosome Location:
Chr.IV: 555726 - 556472 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04109552_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001180358.1 NP_010335.1 1 653 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
glycerol catabolic process
glyceraldehyde-3-phosphate biosynthetic process
triose-phosphate isomerase activity
isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0