This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04131187_s1
See other DNA2 GE Assays ›
Gene Symbol:
DNA2
Gene Name:
Gene Aliases:
WEB2, YHR164C
Chromosome Location:
Chr.VIII: 424610 - 429178 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04131187_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001179295.1 NP_012034.1 1 4449 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

meiotic DNA double-strand break processing
telomere maintenance
DNA double-strand break processing
DNA replication
DNA-dependent DNA replication
lagging strand elongation
DNA repair
cellular response to DNA damage stimulus
metabolic process
DNA duplex unwinding
mitotic DNA replication checkpoint
DNA replication, Okazaki fragment processing
G-quadruplex DNA unwinding
replication fork reversal
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
nucleotide binding
four-way junction DNA binding
DNA binding
DNA helicase activity
RNA binding
catalytic activity
helicase activity
nuclease activity
ATP binding
hydrolase activity
5'-flap endonuclease activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
DNA hairpin binding
protein serine/threonine kinase activator activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
telomeric G-quadruplex DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0