This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04143531_s1
See other GPT2 GE Assays ›
Gene Symbol:
GPT2
Gene Name:
Gene Aliases:
GAT1, YKR067W
Chromosome Location:
Chr.XI: 567918 - 570149 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04143531_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001179857.3 NP_012993.3 1 2154 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid biosynthetic process
CDP-diacylglycerol biosynthetic process
regulation of triglyceride metabolic process
glycerol-3-phosphate O-acyltransferase activity
glycerone-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
sn-1-glycerol-3-phosphate C16:0-DCA-CoA acyl transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0