This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04151557_s1
See other CAT2 GE Assays ›
Gene Symbol:
CAT2
Gene Name:
Gene Aliases:
YCAT, YML042W
Chromosome Location:
Chr.XIII: 192788 - 194800 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04151557_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001182400.1 NP_013670.1 1 1953 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
carnitine metabolic process
carnitine O-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline