This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04151816_s1
See other PIF1 GE Assays ›
Gene Symbol:
PIF1
Gene Name:
Gene Aliases:
TST1, YML061C
Chromosome Location:
Chr.XIII: 148953 - 151532 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04151816_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001182420.1 NP_013650.1 1 2144 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
telomere maintenance via recombination
telomere maintenance
double-strand break repair via break-induced replication
DNA replication
DNA unwinding involved in DNA replication
DNA repair
DNA recombination
cellular response to DNA damage stimulus
translesion synthesis
negative regulation of telomere maintenance via telomerase
DNA duplex unwinding
G-quadruplex DNA unwinding
chromosome organization
negative regulation of telomerase activity
replication fork reversal
ncRNA transcription
nucleotide binding
DNA binding
DNA helicase activity
single-stranded DNA binding
helicase activity
ATP binding
telomerase inhibitor activity
hydrolase activity
telomeric DNA binding
5'-3' DNA helicase activity
G-quadruplex DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0