This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04153333_s1
See other SUB1 GE Assays ›
Gene Symbol:
SUB1
Gene Name:
Gene Aliases:
TSP1, YMR039C
Chromosome Location:
Chr.XIII: 348644 - 349522 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04153333_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001182536.1 NP_013753.1 1 748 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via nonhomologous end joining
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
termination of RNA polymerase II transcription
hyperosmotic response
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of transcription initiation from RNA polymerase II promoter
RNA polymerase III transcriptional preinitiation complex assembly
negative regulation of ascospore formation
DNA binding
chromatin binding
transcription coactivator activity
G-quadruplex DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0