This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04159738_s1
See other RAP1 GE Assays ›
Gene Symbol:
RAP1
Gene Name:
Gene Aliases:
GRC4, GRF1, TBA1, TUF1, YNL216W
Chromosome Location:
Chr.XIV: 241689 - 244172 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04159738_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001183054.1 NP_014183.1 1 2349 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
telomere maintenance
regulation of transcription, DNA-templated
telomere maintenance via telomere lengthening
chromatin silencing at silent mating-type cassette
telomeric heterochromatin assembly
protection from non-homologous end joining at telomere
positive regulation of transcription from RNA polymerase II promoter
establishment of protein localization to telomere
establishment of protein localization to chromatin
G-quadruplex DNA formation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
TFIID-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
DNA binding, bending
TBP-class protein binding
nucleosomal DNA binding
telomeric DNA binding
histone binding
sequence-specific DNA binding
G-quadruplex DNA binding
RNA polymerase II sequence-specific DNA binding transcription factor binding
telomeric G-quadruplex DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0