This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss00952130_m1
See other PPP2R5E GE Assays ›
Gene Symbol:
PPP2R5E
Gene Name:
protein phosphatase 2 regulatory subunit B'epsilon
Gene Aliases:
-
Chromosome Location:
Chr.1: 194505535 - 194653302 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss00952130_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R5E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244062.1 NP_001230991.1 13 - 14 1558 71
GenBank mRNA AK347219.1 - 13 - 14 1558 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline