This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss00961485_g1
See other UBE2E3 GE Assays ›
Gene Symbol:
UBE2E3
Gene Name:
ubiquitin conjugating enzyme E2 E3
Gene Aliases:
-
Chromosome Location:
Chr.15: 86470912 - 86562297 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss00961485_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2E3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021076031.1 XP_020931690.1 797 82
XM_021076032.1 XP_020931691.1 491 82
XM_021076033.1 XP_020931692.1 652 82
XM_021076034.1 XP_020931693.1 366 82
GenBank mRNA AK344952.1 - 2 - 3 571 82
AK350847.1 - 2 - 3 467 82
AK395236.1 - 2 - 3 393 82
AY610396.1 - 1 - 2 219 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0