This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss00963674_m1
See other UBE3A GE Assays ›
Gene Symbol:
UBE3A
Gene Name:
ubiquitin protein ligase E3A
Gene Aliases:
UME3A
Chromosome Location:
Chr.1: 141887179 - 142001595 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss00963674_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243181.2 NP_001230110.2 4 - 5 780 82
NM_001258279.1 NP_001245208.1 2 - 3 594 82
XM_005652466.3 XP_005652523.1 705 82
XM_013999067.2 XP_013854521.1 1022 82
XM_021075796.1 XP_020931455.1 653 82
XM_021075846.1 XP_020931505.1 587 82
XM_021075905.1 XP_020931564.1 528 82
XM_021075965.1 XP_020931624.1 537 82
XM_021076036.1 XP_020931695.1 462 82
XM_021076100.1 XP_020931759.1 653 82
XM_021076243.1 XP_020931902.1 2449 82
XM_021076297.1 XP_020931956.1 609 82
XM_021076371.1 XP_020932030.1 471 82
XM_021076429.1 XP_020932088.1 405 82
XM_021076482.1 XP_020932141.1 418 82
GenBank mRNA AK234494.1 - 4 - 5 780 82
AK348177.1 - 2 - 3 594 82
JQ316194.1 - 2 - 3 168 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001258279.1, XM_005652466.3, XM_013999067.2, XM_021076243.1, XM_021076297.1, XM_021076371.1, XM_021076429.1, XM_021076482.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0