This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss01077055_m1
See other OAZ2 GE Assays ›
Gene Symbol:
OAZ2
Gene Name:
ornithine decarboxylase antizyme 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 107325225 - 107338475 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss01077055_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OAZ2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122995.2 NP_001116467.1 3 - 4 533 124
NM_001301326.1 NP_001288255.1 3 - 4 530 124
GenBank mRNA AK392493.1 - 3 - 4 530 124
BP172930.1 - 3 - 4 533 124
CK454400.1 - 2 - 3 349 124
EU545196.1 - 3 - 4 363 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0