This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss02513702_g1
See other SLC7A7 GE Assays ›
Gene Symbol:
SLC7A7
Gene Name:
solute carrier family 7 member 7
Gene Aliases:
-
Chromosome Location:
Chr.7: 76195774 - 76235406 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss02513702_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110421.1 NP_001103891.1 6 - 7 1130 62
XM_005666264.3 XP_005666321.1 1368 62
XM_005666266.3 XP_005666323.1 1365 62
XM_005666267.3 XP_005666324.1 1356 62
XM_005666269.3 XP_005666326.1 1240 62
XM_013978223.2 XP_013833677.1 3777 62
XM_013978224.2 XP_013833678.1 1359 62
XM_013978225.2 XP_013833679.1 1498 62
XM_013978226.2 XP_013833680.1 1414 62
XM_013978228.2 XP_013833682.1 1231 62
GenBank mRNA AK344424.1 - 5 - 6 752 62
AK345913.1 - 5 - 6 896 62
EU047705.1 - 6 - 7 1130 62
EU390782.2 - 5 - 6 996 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0