This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss02686076_g1
See other SYNCRIP GE Assays ›
Gene Symbol:
SYNCRIP
Gene Name:
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.1: 54563112 - 54598408 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss02686076_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SYNCRIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021089670.1 XP_020945329.1 1189 151
XM_021089676.1 XP_020945335.1 1522 151
XM_021089679.1 XP_020945338.1 1100 151
XM_021089683.1 XP_020945342.1 1522 151
XM_021089690.1 XP_020945349.1 1035 151
XM_021089693.1 XP_020945352.1 1102 151
XM_021089702.1 XP_020945361.1 1037 151
GenBank mRNA AK239681.1 - 2 - 3 226 151
AK344479.1 - 8 - 9 1184 151
AK389965.1 - 8 - 9 1306 151
AK393389.1 - 8 - 9 1186 151
AK395267.1 - 8 - 9 1121 151

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0