This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03210228_m1
See other QKI GE Assays ›
Gene Symbol:
QKI
Gene Name:
QKI, KH domain containing RNA binding
Gene Aliases:
PqkI
Chromosome Location:
Chr.1: 4732575 - 4883386 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03210228_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: QKI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007195.1 NP_001007196.1 4 - 5 547 71
XM_021083630.1 XP_020939289.1 681 71
XM_021083632.1 XP_020939291.1 681 71
XM_021083643.1 XP_020939302.1 692 71
XM_021083654.1 XP_020939313.1 681 71
XM_021083664.1 XP_020939323.1 681 71
XM_021083671.1 XP_020939330.1 682 71
XM_021083682.1 XP_020939341.1 1209 71
XM_021083688.1 XP_020939347.1 681 71
GenBank mRNA AB177988.1 - 4 - 5 547 71
AK390725.1 - 3 - 4 374 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0