This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03375623_u1
See other SLCO1A2 GE Assays ›
Gene Symbol:
SLCO1A2
Gene Name:
solute carrier organic anion transporter family member 1A2
Gene Aliases:
OATP1A2, OATPA
Chromosome Location:
Chr.5: 52020303 - 52133132 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03375623_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256595.1 NP_001243524.1 5 508 96
XM_021091148.1 XP_020946807.1 777 96
XM_021091149.1 XP_020946808.1 769 96
XM_021091150.1 XP_020946809.1 740 96
XM_021091151.1 XP_020946810.1 2527 96
XM_021091152.1 XP_020946811.1 3420 96
XM_021091153.1 XP_020946812.1 2672 96
XM_021091154.1 XP_020946813.1 946 96
XM_021091156.1 XP_020946815.1 2595 96
XM_021091157.1 XP_020946816.1 2450 96
XM_021091158.1 XP_020946817.1 1158 96
XM_021091159.1 XP_020946818.1 920 96
XM_021091160.1 XP_020946819.1 772 96
XM_021091161.1 XP_020946820.1 3415 96
XM_021091162.1 XP_020946821.1 986 96
XM_021091163.1 XP_020946822.1 2446 96
XM_021091164.1 XP_020946823.1 1177 96
GenBank mRNA AF403248.1 - 4 356 96
JQ037799.1 - 5 508 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0