This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03375693_u1
See other GJA1 GE Assays ›
Gene Symbol:
GJA1
Gene Name:
gap junction protein alpha 1
Gene Aliases:
CX43
Chromosome Location:
Chr.1: 40988818 - 41002129 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03375693_u1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244212.1 NP_001231141.1 2 889 91
GenBank mRNA AJ293888.1 - 1 147 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

response to ischemia
spermatogenesis
heart development
response to wounding
negative regulation of cell growth
cellular response to platelet-derived growth factor stimulus
regulation of apoptotic process
bone remodeling
transmembrane transport
bone development
positive regulation of smooth muscle cell chemotaxis
microtubule-based transport
tubulin binding
gap junction hemi-channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline