This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03376505_u1
See other MRPS11 GE Assays ›
Gene Symbol:
MRPS11
Gene Name:
mitochondrial ribosomal protein S11
Gene Aliases:
-
Chromosome Location:
Chr.1: 191310547 - 191321951 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03376505_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPS11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244528.1 NP_001231457.1 5 698 79
XM_005660028.2 XP_005660085.1 120
GenBank mRNA AK234445.1 - 5 744 79
AK240214.1 - 5 698 79
AY550046.1 - 2 45 79
AY609819.1 - 5 458 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline