This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03378516_u1
See other HMOX1 GE Assays ›
Gene Symbol:
HMOX1
Gene Name:
heme oxygenase 1
Gene Aliases:
HSP32
Chromosome Location:
Chr.Unknown: 1954828 - 1963836 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03378516_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004027.1 NP_001004027.1 852 83
GenBank mRNA X60677.1 - 852 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried

Gene Ontology Categories:

Function(s) Process(es)

wound healing involved in inflammatory response
heme oxidation
cellular iron ion homeostasis
apoptotic process
response to oxidative stress
erythrocyte homeostasis
regulation of transcription from RNA polymerase II promoter in response to iron
response to nicotine
heme catabolic process
heme metabolic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation by host of viral transcription
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell proliferation
iron ion homeostasis
cellular response to arsenic-containing substance
cellular response to cadmium ion
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of viral life cycle
heme oxygenase (decyclizing) activity
heme binding
identical protein binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0