This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03379450_u1
See other FABP2 GE Assays ›
Gene Symbol:
FABP2
Gene Name:
fatty acid binding protein 2
Gene Aliases:
I-FABP
Chromosome Location:
Chr.8: 104464342 - 104468059 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03379450_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031780.1 NP_001026950.1 3 351 109
GenBank mRNA AY960624.1 - 3 351 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Gene Ontology Categories:

Function(s) Process(es)

intestinal lipid absorption
long-chain fatty acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-859c6f577-k4jj8:80/100.66.76.192:80.
git-commit: 13ffd1f44fd3b57634547fe5abf56598aa1cdbac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.0-2026.02.07-1.0