This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03379689_u1
See other TPI1 GE Assays ›
Gene Symbol:
TPI1
Gene Name:
triosephosphate isomerase 1
Gene Aliases:
TIM
Chromosome Location:
Chr.5: 63839473 - 63842853 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03379689_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037151.1 NP_001032228.1 3 354 80
GenBank mRNA AB530158.1 - 3 309 80
AK234313.1 - 3 344 80
AK234489.1 - 3 345 80
AK351279.1 - 3 345 80
AK391653.1 - 3 346 80
AK397387.1 - 3 345 80
AK399189.1 - 3 345 80
AK399311.1 - 3 345 80
DQ176425.1 - 3 354 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
methylglyoxal biosynthetic process
glyceraldehyde-3-phosphate biosynthetic process
triose-phosphate isomerase activity
methylglyoxal synthase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0