This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03382854_u1
See other BIRC5 GE Assays ›
Gene Symbol:
BIRC5
Gene Name:
baculoviral IAP repeat containing 5
Gene Aliases:
-
Chromosome Location:
Chr.12: 3747276 - 3755214 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03382854_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BIRC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214141.1 NP_999306.1 3 - 4 349 97
GenBank mRNA AF195781.1 - 3 - 4 349 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
mitotic cytokinesis
protein phosphorylation
apoptotic process
chromosome segregation
mitotic spindle assembly checkpoint
protein complex localization
positive regulation of exit from mitosis
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
cell division
establishment of chromosome localization
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
microtubule binding
zinc ion binding
tubulin binding
protein homodimerization activity
cysteine-type endopeptidase inhibitor activity involved in apoptotic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline